Order Entry
Belgium
ContactUsLinkComponent
Mouse recombinant CD40L, soluble
Mouse recombinant CD40L, soluble
Catalog # ENZOALX850075KI01
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Mouse recombinant CD40L, soluble
Catalog # ENZOALX850075KI01
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 SET
  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Seq:
    02
  • Reconstitution instructions:
    Reconstitute CD40L with 100 µl sterile water and each vial of enhancer with 50 µl of sterile water. Further dilutions should be made with medium containing 5% fetal calf serum or carrier protein
  • Protein/peptide name:
    CD40L, soluble
  • Purity:
    >90% (SDS-PAGE)
  • Concentration:
    CD40L: 0,1 mg/ml after reconstitution
    Enhancer: 1 mg/ml after reconstitution
  • Formulation:
    Lyophilised. Contains PBS.

Specifications

About this item

ELISAFunctional Application: Stimulates the proliferation of mouse B cells and dendritic cells. The activity of CD40L increases 1'000-fold (stimulation in the ng/ml range) in the presence of the cross-linking enhancer (Prod. No. ALX-804-034).

Ordering information: Contents: 10 µg of CD40L, Soluble (mouse) (recombinant) (Prod. No. ALX-522-070) and 2×50 µg of Enhancer for Ligands (Prod. No. ALX-804-034) which increases the biological activity of CD40L at least 1'000-fold.