Order Entry
Belgium
ContactUsLinkComponent
Human recombinant FasL (soluble)
Human recombinant FasL (soluble)
Catalog # ENZOALX850014KI02
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant FasL (soluble)
Catalog # ENZOALX850014KI02
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 SET
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Reconstitution instructions:
    Reconstitute FasL with 100 µl and each vial of enhancer with 50 µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum or a carrier protein
  • Protein/peptide name:
    FasL (soluble)
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    FasL: 0,1 mg/ml after reconstitution
    Enhancer: 1 mg/ml after reconstitution
  • Formulation:
    Lyophilised. Contains PBS.

Specifications

About this item

Kills Fas-sensitive cells at concentrations of >1 ng/ml in the presence of cross-linking enhancer.

Ordering information: Contents: 10 µg of FasL, Soluble (human) (recombinant) (Prod. No. ALX-522-001) and 4×50 µg of Enhancer for Ligands (Prod. No. ALX-804-034) which increases the biological activity (induction of apoptosis) by ~50-fold to a concentration range of 1 to 5 ng/ml.