Human;Mouse recombinant Thy-1 (RLE) (mouse):Fc
ENZOALX522097C050
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Species:HumanMouse
- Seq:02
- Reconstitution instructions:Reconstitute with 50 µl sterile distilled water. Further dilutions should be made with medium containing 5% fetal calf serum or a carrier protein.
- Protein/peptide name:Thy-1 (RLE) (mouse):Fc
- Purity:>90% (SDS-PAGE)
- Concentration:1 mg/ml after reconstitution
- Formulation:Lyophilised. Contains PBS.
Produced in HEK 293 cells. The extracellular domain of mouse Thy-1 (aa 20-131) is fused to the Fc portion of human IgG1. The integrin binding sequence RLD of mouse Thy-1 has been changed to RLE abolishing the binding of integrins.