Order Entry
Belgium
ContactUsLinkComponent
Human;Mouse recombinant Thy-1 (mouse):Fc
Human;Mouse recombinant Thy-1 (mouse):Fc
  ENZOALX522096C050
 :  
Human;Mouse recombinant Thy-1 (mouse):Fc
  ENZOALX522096C050
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
    Mouse
  • Seq:
    02
  • Reconstitution instructions:
    Reconstitute with 50 µl sterile distilled water. Further dilutions should be made with medium containing 5% fetal calf serum or a carrier protein.
  • Protein/peptide name:
    Thy-1 (mouse):Fc
  • Purity:
    >90% (SDS-PAGE)
  • Concentration:
    1 mg/ml after reconstitution
  • Formulation:
    Lyophilised. Contains PBS.

 

 

Produced in HEK 293 cells. The extracellular domain of mouse Thy-1 (aa 20-131) is fused to the Fc portion of human IgG1.