Human recombinant thy-1:Fc
Catalog # ENZOALX522091C050
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Reconstitution instructions:Reconstitute with 50 µl sterile distilled water. Further dilutions should be made with medium containing 5% fetal calf serum or a carrier protein.
- Protein/peptide name:Thy-1:Fc
- Purity:>90% (SDS-PAGE)
- Concentration:1 mg/ml after reconstitution
- Formulation:Lyophilised. Contains PBS.
Specifications
About this item
Produced in HEK 293 cells. The extracellular domain of human Thy-1 (aa 20-130) is fused to the Fc portion of human IgG1.