Human recombinant DcR3:Fc
Catalog # ENZOALX522055C050
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Reconstitution instructions:Reconstitute with 50 µl sterile water.
- Protein/peptide name:DcR3:Fc
- Purity:>90% (SDS-PAGE)
- Concentration:1 mg/ml after reconstitution
- Formulation:Lyophilised. Contains PBS.
Specifications
About this item
Produced in HEK 293 cells. The extracellular domain of human DcR3 (decoy receptor 3) (aa 1-246) is fused to the Fc portion of human IgG1.