Human;Mouse recombinant Fn14 (Mouse):Fc
ENZOALX522036C050
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Species:HumanMouse
- Seq:02
- Biological activity:Inhibits TWEAK-mediated killing of Kym-1 target cells.
- Reconstitution instructions:Reconstitute with 50 µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum.
- Protein/peptide name:Fn14 (mouse):Fc
- Purity:>95% (SDS-PAGE)
- Concentration:1 mg/ml after reconstitution
- Formulation:Lyophilised. Contains PBS.
Produced in HEK 293 cells. The extracellular domain of mouse Fn14 (aa 1-75) is fused to the Fc portion of human IgG1.