Order Entry
Belgium
ContactUsLinkComponent
 
Human Recombinant CD137:Fc
  ENZOALX522031C050
 :  
Human Recombinant CD137:Fc
  ENZOALX522031C050
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Reconstitution instructions:
    Reconstitute with 50 µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum or a carrier protein.
  • Protein/peptide name:
    CD137:Fc
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    1 mg/ml after reconstitution
  • Formulation:
    Lyophilised. Contains PBS.

 

 

Produced in HEK 293 cells. The cysteine-rich region of human CD137 (4-1BB) (aa 1-186) is fused to the Fc portion of human IgG1.