Human recombinant BCMA:Fc
ENZOALX522026C050
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Biological activity:Blocks the binding of BAFF and APRIL to their receptors BCMA and TACI, inhibiting BAFF- and APRIL-mediated B cell activation.
- Reconstitution instructions:Reconstitute with 50 µl of sterile water. Further dilutions should be made with medium containing 5% fetal calf serum or a carrier protein.
- Protein/peptide name:BCMA:Fc
- Purity:>95% (SDS-PAGE)
- Concentration:1 mg/ml after reconstitution
- Formulation:Lyophilised. Contains PBS.
Produced in HEK 293 cells. The extracellular domain of human BCMA (aa 2-54) is fused at the C-terminus to the Fc portion of human IgG1.