Human recombinant TNF-R2:Fc
ENZOALX522014C050
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Reconstitution instructions:Reconstitute with 50 µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum or a carrier protein.
- Protein/peptide name:TNF-R2:Fc
- Purity:>95% (SDS-PAGE)
- Concentration:1 mg/ml after reconstitution
- Formulation:Lyophilised. Contains PBS.
Produced in HEK 293 cells. The extracellular domain of human TNF-R2 (aa 1-257) is fused to the Fc portion of human IgG1.