Human recombinant TNF alpha (soluble) (from E. coli)
Catalog # ENZOALX522008C050
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Biological activity:Exerts its biological activity in a concentration range of 0,1 - 1 ng/ml (WEHI 164 cells).
- Reconstitution instructions:Reconstitute with 50 µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum.
- Protein/peptide name:TNF alpha (soluble)
- Purity:>95% (SDS-PAGE)
- Concentration:1 mg/ml after reconstitution
- Formulation:Lyophilised. Contains PBS.
Specifications
About this item
Produced in E. coli. The extracellular domain of human TNF-α (tumor necrosis factor-α) (aa 85-223) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG®-tag.