Order Entry
Belgium
ContactUsLinkComponent
 
Human recombinant TRAILR2:Fc
  ENZOALX522005C050
 :  
Human recombinant TRAILR2:Fc
  ENZOALX522005C050
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Biological activity:
    Inhibits human soluble TRAIL-induced apoptosis in a concentration range of 0,5 - 10 µg/ml.
  • Reconstitution instructions:
    Reconstitute with 50 µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum or a carrier protein.
  • Protein/peptide name:
    TRAILR2:Fc
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    1 mg/ml after reconstitution
  • Formulation:
    Lyophilised. Contains PBS. Resuspend in 50 µl sterile water.

 

 

Produced in HEK 293 cells. The extracellular domain of human TRAIL-R2 (DR5) (aa 52-212) is fused to the Fc portion of human IgG1.