Order Entry
Belgium
ContactUsLinkComponent
 
Human recombinant TRAIL (soluble) (from E. coli)
 
Human recombinant TRAIL (soluble) (from E. coli)

 

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Biological activity:
    Induces apoptosis in a concentration range of 1 - 100 ng/ml if applied with the cross-linking enhancer (Prod. No. ALX-804-034); see also TRAIL, Soluble (human) (recombinant) Set. Note: rhsTRAIL does not significantly induce apoptosis
  • Reconstitution instructions:
    Reconstitute with 100 µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum.
  • Protein/peptide name:
    TRAIL (soluble)
  • Purity:
    >90% (SDS-PAGE)
  • Concentration:
    0,1 mg/ml after reconstitution
  • Formulation:
    Lyophilised. Contains PBS.

 

Frequently Bought Together