Order Entry
Belgium
ContactUsLinkComponent
Microcystis aeruginosa [D-Asp3]microcystin-LR
Microcystis aeruginosa [D-Asp3]microcystin-LR
Supplier:  Enzo Life Sciences
Certificates
undefined
Microcystis aeruginosa [D-Asp3]microcystin-LR
Supplier:  Enzo Life Sciences

Specifications

  • Species:
    Microcystis aeruginosa
  • Protein/peptide name:
    [D-Asp3]microcystin-LR
  • Purity:
    >95% (HPLC)

Specifications

Frequently Bought Together