Bovine native Ganglioside GT1b . trisodium salt (from Brain)
Supplier: Enzo Life Sciences
Certificates
About this item
Prevents neurotoxicity of glutamate on neuronal cells.
Specifications
- Protein/peptide type:Native
- Source:Brain
- Species:Bovine
- Protein/peptide name:Ganglioside GT1b . trisodium salt
- Purity:>98% (TLC)
Specifications
Frequently Bought Together