Z-Phe-Arg-pNA
Catalog # ENZOALX260130M005
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:5 mg
- Seq:02
- Protein/peptide name:Z-Phe-Arg-pNA
- Purity:__
- Formulation:Lyophilized
Specifications
About this item
Cathepsin substrate
Chromogenic substrate for Cathepsins B, K, L, and S, papain, trypsin, and plasma kallikrein. This substrate is likely to be cleaved by other cathepsins (see Z-Phe-Arg-AMC). Release of pNA is monitored at 405 to 410 nm (pNA ε410=8800 M-1cm-1). Useful for inhibitor screening and kinetic analysis.