Mca-Pro-Leu-Ala-Nva-Dap(Dnp)-Ala-Arg-NH2
Catalog # ENZOALX260123M001
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:1 mg
- Seq:02
- Protein/peptide name:Mca-Pro-Leu-Ala-Nva-Dap(Dnp)-Ala-Arg-NH2
- Purity:>95% (HPLC)
Specifications
About this item
MMP-2 substrate
Fluorogenic substrate for MMP-2.