Ac-VDVAD-AFC
Caspase-2 substrate
Fluorogenic substrate for caspase-2; similar to Ac-VDVAD-AMC but the AFC fluorophore has a greater Stokes’ shift upon cleavage. VDVAD has been found to be a preferred cleavage site for caspase-2 (Ich-1L). Increases in Ac-VDVAD-AFC cleavage correlated with losses in procaspase-2 (conversion to active caspase-2) in human neuroblastoma lines induced to apoptosis with C2-ceramide and NO.
- Protein/peptide name:Ac-VDVAD-AFC
- Purity:>98% (HPLC)
Frequently Bought Together