ContactUsLinkComponent
Ac-VAD-AFC
Ac-VAD-AFC
Catalog # ENZOALX260109M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Ac-VAD-AFC
Catalog # ENZOALX260109M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Pk:
    5 mg
  • Seq:
    02
  • Protein/peptide name:
    Ac-VAD-AFC
Specifications
About this item
Caspase substrate
Fluorogenic substrate for all caspases.