Order Entry
Belgium
ContactUsLinkComponent
Human recombinant Annexin V (from E. coli), RPE (R-Phycoerythrin)
Human recombinant Annexin V (from E. coli), RPE (R-Phycoerythrin)
Supplier:  ENZO LIFE SCIENCES
Certificates
Human recombinant Annexin V (from E. coli), RPE (R-Phycoerythrin)
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Conjugation:
    RPE (R-Phycoerythrin)
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Protein/peptide name:
    Annexin V
  • Purity:
    >98% (SDS-PAGE, HPLC)
  • Formulation:
    Liquid. In 50 mM TRIS, pH 7,4, containing 100 mM sodium chloride, 1% BSA and 0,02% sodium azide.

Specifications

Frequently Bought Together