Human recombinant Annexin V (from E. coli), FITC (Fluorescein Isothiocyanate)
Produced in E. coli.
FITC-labelled recombinant human annexin V shows bright green fluorescence (Ex(max): 488 nm; Em(max): 530 nm).
: Quantity: 20 tests/100 µl; 100 tests/500 µl; 300 tests/1500 µl.
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Biological activity:Exhibits anti-phospholipase activity.
- Protein/peptide name:Annexin V
- Purity:>98% (SDS-PAGE, HPLC)
- Formulation:Liquid. In 50 mM TRIS, pH 7,4, containing 100 mM sodium chloride, 1% BSA and 0,02% sodium azide.
Frequently Bought Together