Order Entry
Belgium
ContactUsLinkComponent
Bovine native Calmodulin (from Brain), Biotin
Bovine native Calmodulin (from Brain), Biotin
Catalog # ENZOALX209001C050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Bovine native Calmodulin (from Brain), Biotin
Catalog # ENZOALX209001C050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Conjugation:
    Biotin
  • Protein/peptide type:
    Native
  • Source:
    Brain
  • Species:
    Bovine
  • Seq:
    02
  • Protein/peptide name:
    Calmodulin
  • Concentration:
    0,5 mg/ml
  • Formulation:
    Liquid. In PBS containing 0,1% BSA and 0,02% sodium azide.
  • Grade:
    High purity

Specifications

About this item

Isolated from bovine brain.