Mouse Recombinant ACRP30headless
ENZOALX203003C010
:
- Pk:10 µG
- Protein/peptide type:Recombinant
- Species:Mouse
- Seq:02
- Reconstitution instructions:Reconstitute with 100 µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum or a carrier protein.
- Protein/peptide name:ACRP30 headless
- Purity:>90% (SDS-PAGE)
- Concentration:0,1 mg/ml after reconstitution.
- Formulation:Lyophilised. Contains PBS.
Produced in HEK 293 cells. The collagen domain of mouse ACRP30 (aa 18-111) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG®-tag.