Human;Rat recombinant COMP:Angiopoietin-1 (from CHO cells)
Supplier: Enzo Life Sciences
Certificates
About this item
Soluble, stable and more potent Ang-1 variant.
Produced in CHO cells. The fibrinogen-like domain (aa 284-498) of human Ang-1 (angiopoietin-1) is fused at the N-terminus to the coiled-coil domain of rat COMP (cartilage oligomeric matrix protein) and to a FLAG®-tag.
Soluble, stable and more potent Ang-1 variant.
Specifications
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:HumanRat
- Biological activity:Induces phosphorylation of Tie-2 in primary cultured human and mouse vascular endothelial cells. More potent than native Ang-1.
- Protein/peptide name:COMP:Angiopoietin-1
- Purity:>90% (SDS-PAGE)
- Concentration:~0,3 mg/ml
- Formulation:Liquid. 0,2 µM-filtered solution in PBS containing 0,01% CHAPS.
Specifications
Frequently Bought Together