Human recombinant NF-kappaB (p50) (from insect cells (Sf9)), HIS tag
Catalog # ENZOALX201285C002
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:2 µG
- Protein/peptide type:Recombinant
- Source:Insect cells (Sf9)
- Species:Human
- Seq:02
- Tag sequence:HIS Tag
- Protein/peptide name:NF-kappaB (p50)
- Purity:>95% (SDS-PAGE)
- Concentration:0,2 mg/ml
- Formulation:Liquid. In 50 mM TRIS-HCl, pH 7,5, containing 100 mM sodium chloride, 0,2% NP-40, 50 - 100 mM imidazole and 10% glycerol.
Specifications
About this item
Produced in SF21 cells. Fused to a His-tag.