Human recombinant procathepsin K (from E. coli)
Catalog # ENZOALX201239C010
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Protein/peptide name:Procathepsin K
- Purity:>95% (SDS-PAGE)
- Concentration:150 μg/ml
- Formulation:Liquid. In 25 mM TRIS, pH 8,0, containing 500 mM sodium chloride.
Specifications
About this item
Produced in E. coli (aa 19-329). Methionine was introduced at aa 18 to create a new N-terminal sequence (MEEIL).
Member of the papain cysteine proteinase family identified as the predominant proteinase responsible for the resorption of the bone matrix.