Human recombinant NMNAT3 (from E. coli)
Catalogus # ENZOALX201238C100
Leverancier: ENZO LIFE SCIENCES
CAS-nummer:
Specificaties
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Protein/peptide name:NMNAT3
- Purity:>95% (SDS-PAGE)
- Concentration:5 mg/ml
- Formulation:Liquid. In 50 mM TRIS-HCl, pH 8,0, containing 300 mM sodium chloride, 1 mM DTT and 10% glycerol.
Specificaties
Over dit artikel
Produced in E. coli. Full length human NMNAT3 is fused at the N-terminus to a His-tag.