Order Entry
Belgium
ContactUsLinkComponent
Recombinant DnaK (1-638) (from E. coli)
Recombinant DnaK (1-638) (from E. coli)
Catalog # ENZOALX201217C100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Recombinant DnaK (1-638) (from E. coli)
Catalog # ENZOALX201217C100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Seq:
    02
  • Protein/peptide name:
    DnaK (1-638)
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    1 mg/ml
  • Formulation:
    Liquid. In 20 mM TRIS buffer, pH 7,5, containing 100 mM sodium chloride, 5 mM DTT and 10% glycerol.

Specifications

About this item

Recombinant full length DnaK produced in E. coli.