Order Entry
Belgium
ContactUsLinkComponent
Human recombinant SET7/9 (from E. coli)
Human recombinant SET7/9 (from E. coli)
Catalog # ENZOALX201178C100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant SET7/9 (from E. coli)
Catalog # ENZOALX201178C100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    SET7/9
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    1 mg/ml
  • Formulation:
    Liquid. In 50 mM TRIS-HCl, pH 7,5, containing 0,2 M sodium chloride, 5 mM DTT and 20% glycerol.

Specifications

About this item

Produced in E. coli.