Order Entry
Belgium
ContactUsLinkComponent
Mouse recombinant PARP2 (from insect cells (Sf9))
Mouse recombinant PARP2 (from insect cells (Sf9))
Catalog # ENZOALX201064C020
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Mouse recombinant PARP2 (from insect cells (Sf9))
Catalog # ENZOALX201064C020
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    20 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Insect cells (Sf9)
  • Species:
    Mouse
  • Seq:
    02
  • Protein/peptide name:
    PARP2
  • Purity:
    >98% (SDS-PAGE)
  • Concentration:
    ~1 mg/ml
  • Formulation:
    Liquid. In 50 mM TRIS-HCl, pH 7,5, containing 1 mM DTT, 0,5 mM EDTA, 0,5 mM PMSF and 20% glycerol.
  • Grade:
    High purity
  • Purification:
    Affinity purified

Specifications

About this item

Produced in Sf9 cells (full length).