Order Entry
Belgium
ContactUsLinkComponent
Human native myelin basic protein (from Brain)
Human native myelin basic protein (from Brain)
Catalog # ENZOALX200606M001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human native myelin basic protein (from Brain)
Catalog # ENZOALX200606M001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Native
  • Source:
    Brain
  • Species:
    Human
  • Seq:
    02
  • Reconstitution instructions:
    Reconstitute with distilled water.
  • Protein/peptide name:
    Myelin basic protein
  • Purity:
    >95% (SDS-PAGE; Immunoblot)
  • Concentration:
    1 mg/ml after reconstitution.
  • Formulation:
    Lyophilised. Essentially salt-free.
  • Grade:
    High purity

Specifications

About this item

Purified from human brain by method of Määttä et al..