Human granzyme A
Catalog # ENZOALX200605C010
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:10 µG
- Species:Human
- Seq:02
- Protein/peptide name:Granzyme A
- Concentration:2,76 mg/ml
- Formulation:Liquid. In ~0,15 M sodium chloride.
- Purification:Purified by differential solubilisation and cation exchange chromatography. Dialysed against 0,15 M sodium chloride and concentrated.
Specifications
About this item
Human cytotoxic granules of NK-92 cells.