Order Entry
Belgium
ContactUsLinkComponent
Pam3Cys-Ser-(Lys)4 . trihydrochloride
Pam3Cys-Ser-(Lys)4 . trihydrochloride
Catalog # ENZOALX165066M002
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Pam3Cys-Ser-(Lys)4 . trihydrochloride
Catalog # ENZOALX165066M002
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    2 mg
  • Seq:
    02
  • Reconstitution instructions:
    Reconstitute in 2 ml distilled water. Use a homogeniser or sonicator to prepare a homogenous solution.
  • Protein/peptide name:
    Pam3Cys-Ser-(Lys)4 . trihydrochloride
  • Concentration:
    1 mg/ml after reconstitution.
  • Formulation:
    Lyophilised.

Specifications

About this item

Selective agonist of TLR1/TLR2 complex

Selective agonist of Toll-like receptor 1 (TLR1) complexed with TLR2. Cell permeable, water soluble synthetic cationic lipohexapeptide analog of the immunologically active N-terminal portion of bacterial lipoprotein that potently activates monocytes and macrophages. Activates/represses an array of at least 140 genes partly involved in signal transduction and regulation of the immune response.