Order Entry
Belgium
ContactUsLinkComponent
Phospholipase A2 activating peptide
Phospholipase A2 activating peptide
Catalog # ENZOALX165007MC01
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Phospholipase A2 activating peptide
Catalog # ENZOALX165007MC01
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    0,1 mg
  • Seq:
    02
  • Protein/peptide name:
    Phospholipase A2 activating Peptide
  • Purity:
    >97%

Specifications

About this item

Peptide fragment from phospholipase A2 (PLA2) activating protein (PLAP) that activates phospholipase A2 in a dose-dependent manner (10-fold at 1 µg/ml).

  • Member of the beta-transducin (Gbeta) superfamily

The amino acid sequence bears considerable homology to melittin.