Order Entry
Belgium
ContactUsLinkComponent
Bovine;Human;Mouse;Rat Orexin A
Bovine;Human;Mouse;Rat Orexin A
Supplier:  ENZO LIFE SCIENCES
Certificates
Bovine;Human;Mouse;Rat Orexin A
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Species:
    Human
    Bovine
    Rat
    Mouse
  • Protein/peptide name:
    Orexin A
  • Purity:
    >98% (HPLC)
  • Formulation:
    Lyophilised

Specifications

Frequently Bought Together