Order Entry
Belgium
ContactUsLinkComponent
Human recombinant HOP (from E. coli)
Human recombinant HOP (from E. coli)
Supplier:  ENZO LIFE SCIENCES
Certificates
Human recombinant HOP (from E. coli)
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Protein/peptide name:
    HOP
  • Purity:
    >90% (SDS-PAGE; Western blot)
  • Formulation:
    Liquid. In Dulbecco's PBS.
  • Purification:
    Purified by multi-step chromatography.

Specifications

Frequently Bought Together