Human recombinant HSF1 (from E. coli), HIS tag
Catalog # ENZOADISPP900F
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:200 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:04
- Tag sequence:HIS Tag
- Protein/peptide name:HSF1
- Purity:-
- Formulation:-
- Purification:-
Specifications
About this item
Produced in E. coli. Human HSF1 is fused at the N-terminus to a His-tag.