Order Entry
Belgium
ContactUsLinkComponent
Human recombinant Calnexin (lumenal domain) (from E. coli)
Human recombinant Calnexin (lumenal domain) (from E. coli)
Supplier:  ENZO LIFE SCIENCES
Certificates
Human recombinant Calnexin (lumenal domain) (from E. coli)
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Protein/peptide name:
    Calnexin (lumenal domain)
  • Purity:
  • Formulation:
  • Purification:

Specifications

Frequently Bought Together