Order Entry
Belgium
ContactUsLinkComponent
Chinook salmon recombinant HSP70/HSP72 (from E. coli)
Chinook salmon recombinant HSP70/HSP72 (from E. coli)
Supplier:  ENZO LIFE SCIENCES
Certificates
Chinook salmon recombinant HSP70/HSP72 (from E. coli)
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Chinook salmon
  • Protein/peptide name:
    HSP70/HSP72
  • Purity:
  • Formulation:
  • Purification:

Specifications

Frequently Bought Together