Bovine Recombinant HSC70 (from E. coli)
Supplier: Enzo Life Sciences
Certificates
About this item
Produced in E. coli.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Bovine
- Protein synonyms:HSP73
- Protein/peptide name:HSC70
- Purity:-
- Formulation:-
- Purification:-
Specifications
Frequently Bought Together