Recombinant DnaK (from E. coli)
Supplier: ENZO LIFE SCIENCES
New Product
Certificates
About this item
Produced in E. coli.
Specifications
- Source:
- Species:
- Environmentally Preferable:
Specifications
Frequently Bought Together
Produced in E. coli.