Human recombinant Calreticulin (from E. coli), N-terminal His-tag
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Produced in E. coli. Human calreticulin is fused at the N-terminus to a His-tag.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:N-terminal His-tag
- Protein/peptide name:Calreticulin
- Purity:>90% (SDS-PAGE; Western blot) (~80% of full length protein)
- Formulation:Liquid, in 50 mM TRIS, pH 7,5, containing 150 mM sodium chloride, 5 mM calcium chloride and 5% glycerol
- Purification:Purified by multi-step chromatography
Specifications
Frequently Bought Together