Human recombinant HSP40/Hdj2 (from E. coli)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Produced in E. coli.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Protein/peptide name:HSP40/Hdj2
- Purity:>90% (SDS-PAGE; Western blot)
- Formulation:Liquid, in TBS containing 1 mM DTT
- Purification:Purified by multi-step chromatography
Specifications
Frequently Bought Together