Mouse recombinant Mn SOD (from E. coli), HIS Tag
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Produced in E.coli. Mouse Mn SOD is fused at the N-terminus to a His-tag.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Tag sequence:HIS Tag
- Protein/peptide name:Mn SOD
- Purity:>90% (SDS-PAGE; Western blot)
- Formulation:Liquid, In 50 mM TRIS-hydrogen chloride, pH 7,5, containing 1,5% glycerol and 80 mM sodium chloride
- Purification:Purified by multi-step chromatography
Specifications
Frequently Bought Together