Human recombinant PKA catalytic beta (from insect cells (Sf9)), GST Tag
Catalog # ENZOADIPPK448Z
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:5 µG
- Protein/peptide type:Recombinant
- Source:Insect cells (Sf9)
- Species:Human
- Seq:02
- Tag sequence:GST Tag
- Protein/peptide name:PKA Catalytic beta
- Purity:>90% (Densitometry)
- Formulation:Liquid, In TRIS-hydrogen chloride, pH 7,5, containing 150 mM sodium chloride, 0,25 mM DTT, 0,1 mM EGTA, 0,1 mM EDTA, 0,1 mM PMSF, and 25% glycerol
- Purification:Purified by multi-step chromatography
Specifications
About this item
Produced in Sf9 cells using an N-terminal GST-tag.