EGTA (2X) Buffer, Enzo Life Sciences
EGTA buffer, 2X, 1 mL
Catalog # ENZOBMLKI1360001
Supplier: Enzo Life Sciences
EGTA (2X) Buffer, Enzo Life Sciences
Catalog # ENZOBMLKI1360001
Supplier: Enzo Life Sciences
CAS Number:
Aller à :
Product Details & Documents
Spécifications
Spécifications
- Concentration:2X
- Pk:1 mL
- Description:EGTA buffer
- Cat. No.:ENZOBMLKI1360001
- Size:1 ml
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



