ContactUsLinkComponent
HDAC assay buffer II
HDAC assay buffer II
Catalog # ENZOBMLKI4220020
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
HDAC assay buffer II
Catalog # ENZOBMLKI4220020
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Pk:
    20 mL
  • Description:
    HDAC assay buffer II
  • Seq:
    02
Specifications
About this item
50 mM Tris/Cl, pH 8.0, 137 mM NaCl, 2.7 mM KCl, 1 mM MgCl2, 1 mg/ml BSA.