Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Proteasome assay buffer

Proteasome assay buffer, 20 mL
Catalog # ENZOBMLKI3400020
Supplier:   Enzo Life Sciences
Proteasome assay buffer
Catalog # ENZOBMLKI3400020
Supplier:  Enzo Life Sciences
CAS Number:  
Proteasome assay buffer

Proteasome assay buffer, 20 mL

CAS Number:  
Price
52,80 €
Log in for availability
1 * 20 mL
Product Details & Documents

Assay buffer from the 20S Proteasome Assay Kit for Drug Discovery.

Specifications

Specifications

  • Pk:
    20 mL
  • Description:
    Proteasome assay buffer

Specifications