Order Entry
Belgium
Orders LinkContactUsLinkComponent
Permeabilisation, (buffer)

Permeabilisation, (buffer)

Permeabilization, (buffer), 100 mL
Catalog # ENZOADI950012100
Supplier:   Enzo Life Sciences
Permeabilisation, (buffer)
Catalog # ENZOADI950012100
Supplier:  Enzo Life Sciences
CAS Number:  
Permeabilisation, (buffer)

Permeabilization, (buffer), 100 mL

CAS Number:  
Price
160,00 €
Log in for availability
1 * 100 mL
Product Details & Documents
Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    Permeabilization, (buffer)
  • Cat. No.:
    ENZOADI950012100
  • Formulation:
    Liquid. In PBS, pH 7,2, containing 0,1% Saponin and 0,09% sodium azide.

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.