Order Entry
Belgium
ContactUsLinkComponent
 
Cell lysis buffer 4, (100 ml), Enzo Life Sciences
  ENZOADI801339
 :  
Cell lysis buffer 4, (100 ml), Enzo Life Sciences
  ENZOADI801339
 :  

 

  • Pk:
    100 mL
  • Description:
    Cell lysis buffer 4
  • Seq:
    02
  • Formulation:
    Liquid, 100 mm PBS, pH 7,3, containing 150 mm sodium chloride, 1 mm EDTA, 0,5% Triton X-100 and 0,005% Tween 20